Learn More
Electron Microscopy Sciences R 1/2 Holey Carbon Films on Grids, Grid: Copper. Mesh: 400., 50/PK
SDP
Catalog No. 502091570
Encompass_Preferred
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 1 µm. Space between holes is 1 µm. Center to center is 2 µm. Smaller holes are designed for magnifications of 50,000x or higher. Grid: Copper. Mesh: 400 .
Catalog No. 50-209-1570 Supplier Electron Microscopy Sciences Supplier No. Q450CR12
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 1 µm. Space between holes is 1 µm. Center to center is 2 µm. Smaller holes are designed for magnifications of 50,000x or higher. Grid: Copper. Mesh: 400 .