Learn More
Electron Microscopy Sciences Quantifoil® R 2/1 200 Mesh, Ni
SDP
Catalog No. NC3761425
Encompass_Preferred
Quantifoil® R 2/1 200 Mesh, Ni. Has more open area than R 2/2. It is used when focusing is carried out on the edge of a hole burnt in the ice in a neighboring hole instead of on the foil adjacent to the hole. Hole Size: 2 µm, Period: 3 µm
Catalog No. NC3761425 Supplier Electron Microscopy Sciences Supplier No. Q2100NR1
Quantifoil® R 2/1 200 Mesh, Ni. Has more open area than R 2/2. It is used when focusing is carried out on the edge of a hole burnt in the ice in a neighboring hole instead of on the foil adjacent to the hole. Hole Size: 2 µm, Period: 3 µm