Learn More
Electron Microscopy Sciences QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil.
SDP
Catalog No. NC3371920
Encompass_Preferred
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement.
Catalog No. NC3371920 Supplier Electron Microscopy Sciences Supplier No. Q2100CR2S
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement.