Learn More
Electron Microscopy Sciences Perforated support foil with pre-defined holes has advantages in electron microscopy compared with conventional holey film. 3 µm holes with 8 µm period.
SDP
Catalog No. NC3760730
Encompass
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 3 µm. Space between holes is 5 µm. Center to center is 8 µm.
Catalog No. NC3760730 Supplier Electron Microscopy Sciences Supplier No. Q250CR5
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 3 µm. Space between holes is 5 µm. Center to center is 8 µm.