Learn More
Electron Microscopy Sciences R 2/2 Holey Carbon Films, 100/PK
SDP
Catalog No. NC1647268
Encompass_Preferred
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 2 µm. Space between holes is 2 µm. Center to center is 4 µm. Types with 2 µm holes are recommended for magnifications between 30.000x and 40.000x.
Catalog No. NC1647268 Supplier Electron Microscopy Sciences Supplier No. LFH2100AR2
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 2 µm. Space between holes is 2 µm. Center to center is 4 µm. Types with 2 µm holes are recommended for magnifications between 30.000x and 40.000x.