Learn More
Electron Microscopy Sciences R 3/3 Holey Carbon Films on Grids, Grid: Copper. Mesh: 300., 25/PK
SDP
Catalog No. NC1906472
Encompass_Preferred
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 3 µm. Space between holes is 3 µm. Center to center is 6 µm.
Catalog No. NC1906472 Supplier Electron Microscopy Sciences Supplier No. Q325CR3
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 3 µm. Space between holes is 3 µm. Center to center is 6 µm.