Learn More
Electron Microscopy Sciences Quantifoil Holey Carbon Film S 7/2 on Copper 400 mesh Grid
SDP
Catalog No. 501902484
Encompass_Preferred
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement. It has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) microscopy when compared with conventional holey film.
QUANTIFOIL with square holes and relatively narrow bars can be used in EM to support a thin carbon film, which by itself is too fragile to span a grid square. Alternatively, this holey film can directly support an object that is larger than the holes.
Catalog No. 50-190-2484 Supplier Electron Microscopy Sciences Supplier No. Q450CS7
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement. It has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) microscopy when compared with conventional holey film.
QUANTIFOIL with square holes and relatively narrow bars can be used in EM to support a thin carbon film, which by itself is too fragile to span a grid square. Alternatively, this holey film can directly support an object that is larger than the holes.