Learn More
Structure Probe Inc QUANTIFOIL ULTRAUFOILR 1.2/1.3
SDP
Catalog No. NC2137329
Encompass
Quantifoil UltrAuFoil R 1.2/1.3 Holey Gold Films
This item is not returnable. View return policy
Catalog No. NC2137329 Supplier Structure Probe Inc Supplier No. 4976GFA
Quantifoil UltrAuFoil R 1.2/1.3 Holey Gold Films