Learn More
Electron Microscopy Sciences Quantifoil Holey Carbon Film Multi A on Nickel 400 mesh Grid
SDP
Supplier:  Electron Microscopy Sciences Q450NMA
Encompass_Preferred
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement. It has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) microscopy when compared with conventional holey film.
QUANTIFOIL Multi A is a holey film, in which a pattern of various hole sizes, shapes and arrangements is repeated. In addition to circular holes, the pattern includes oval-shaped ones which appear round at high tilt angles (70°). The diameters of the round holes are about 1, 1.4, and 2 µm, and the bar widths range from 0.5 to 4 µm. The oval holes in the foil have a dimension of 8 x 2 µm and 4 x 1 µm
Catalog No. 50-190-2503