Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Temperature Validation Kit

Catalog No. QS0517
Encompass_Preferred
Change view
Click to view available options
No. of Places:
96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Places
QS0517 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QS0517 Supplier Invitrogen™ Supplier No. QS0517
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The QuantiGene Incubator Temperature Validation Kit is designed to precisely measure the temperatures in the incubators used in QuantiGene Singleplex and QuantiGene Plex assays.

The kit uses a National Institute of Standards and Technology (NIST)-calibrated thermometer to enable users to assess the accuracy of the temperature of ovens or incubators. The kit can be used to monitor incubators used for QuantiGene assays to ensure adherence to recommended incubation conditions.

What you get
The QuantiGene Incubator Temperature Validation Kit contains a QuantiGene mock hybridization 96-well plate, an NIST-calibrated thermometer and thermocouple probe, and a procedure for validating a static or shaking incubator.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 NIST-calibrated digital thermometer
• 1 Sealed hybridization plate
For Use With (Equipment) QuantiGene™ Incubator
No. of Places 96
Type Temperature Validation Kit
Product Line QuantiGene
Quantity 1 kit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.