Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Lysis Mixture

Catalog No. QP0522
Encompass_Preferred
Change view
Click to view available options
Quantity:
12 mL
36 mL
120 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QP0522 12 mL
QP0523 36 mL
QP0524 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. QP0522 Supplier Invitrogen™ Supplier No. QP0522
Add to Cart
Add to Cart

QuantiGene Lysis Mixture is used to lyse cells from cell culture or whole blood, in order to release target RNA to be quantified with QuantiGene assays.

QuantiGene Lysis Mixture is used to lyse cells from cell culture or whole blood, in order to release target RNA to be quantified with QuantiGene assays. QuantiGene Lysis Mixture is one of two components in the QuantiGene Sample Processing kits for cells and whole blood, and is intended to supplement these kits when additional volume is required.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Multi-Gene Expression
Product Line QuantiGene
Product Type Cell Lysis Buffer
Quantity 12 mL
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.