Learn More
Invitrogen™ QuantiGene™ Plex Assay Kit
Description
The QuantiGene Plex Assay Kit contains the generic reagents, buffers, and plates necessary to perform the assay in 96-well plates and must be used with a target-specific QuantiGene Plex panel (sold separately).
The QuantiGene Plex Assay is a gene expression assay that enables multiplexed measurement of up to 80 RNA transcript targets. The assay uses target-specific probes to capture the RNA of interest, and branched DNA technology then amplifies fluorescent signals which are read with a Luminex xMAP instrument.
Features of the QuantiGene Plex Assay include:
- Difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
- True multiplexing—measure up to 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
- Standardized platform—uses 96 or 384-well plate formats compatible with the INTELLIFLEX, Luminex 200, FlexMap3D, and MagPix systems
- Simple workflow—employs an ELISA-like workflow for direct hybridization of transcripts to beads and transcript labeling
- Large inventory of validated genes—select from over 15,000 genes to create pathway and disease-themed panels
- Rapid turnaround for custom panels—if we don't have your gene(s), we can create a custom panel within two weeks
Technology
QuantiGene assays utilize branched DNA (bDNA) technology that has been used for decades in the VERSANT™ diagnostic assays for HIV-1, HCV, and HBV. First, cells are lysed or tissue samples are homogenized to release the target RNA. Second, the oligonucleotide probe set is incubated with the target RNA and Luminex xMAP bead set overnight. During this incubation, the probes cooperatively hybridize to the target and capture probes bound to each bead, capturing the target RNA. Third, signal amplification is performed via sequential hybridization of the bDNA preamplifier, amplifier, and label-probe molecules to the target. Addition of streptavidin phycoerythrin generates a luminescent signal directly proportional to the amount of target mRNA present in the sample.
Applications
QuantiGene Plex Assay applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency, prospective and/or retrospective analysis of clinical samples, miRNA profiling, microarray validation, predictive toxicology, and detection of translocations and fusion genes.
Specifications
Specifications
| Format | 96-well |
| For Use With (Application) | QuantiGene Plex Panel |
| For Use With (Equipment) | Luminex instrument |
| Content And Storage | • 350 μL Proteinase K, –20°C Note: Prepared 1x Wash Buffers can be stored for 1 week protected from light at 4°C. |
| Product Line | QuantiGene Plex |
| Product Type | Multiplex Assay Kit |
| Quantity | 10 Plates |
| Shipping Condition | Blue Ice |
| Sample Type | Purified RNA, cell/blood/FFPE lysates, tissue homogenates |
| Detection Method | Fluorescence |
| Show More |
Frequently Asked Questions (FAQs)
No, samples in TRIzol solution cannot be run in the QuantiGene Plex assay. However, RNA purified from these samples can be run in the QuantiGene Plex assay.
Although the kits were not originally validated for bacteria, we have guidelines for preparing bacterial samples for QuantiGene Plex assays: http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/preparation-bacterial-homogenates-quantigene-plex-assays-protocol.pdf
Yes, please reach out to your Technical Sales Specialist for more information or fill out this form: https://www.thermofisher.com/us/en/home/global/forms/life-science/request-quote-quantigene-assays.html
Under the hybridization conditions that are used for the QuantiGene Plex assay, genomic DNA remains double-stranded. Therefore, it is not possible for the probes to bind to DNA. The signal from the probe sets comes only from RNA.
We only recommend the use of QuantiGene Lysis Mixture for preparing cell and blood lysates. The Lysis Mixture has been optimized to provide stability of RNA in the lysates. In addition, QuantiGene Lysis Mixture provides the appropriate buffer conditions for the overnight hybridization.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.