Learn More
Invitrogen™ QuantiGene™ Plex Hybridization Plates
Description
QuantiGene Plex Hybridization Plates are used for the overnight hybridization step of the QuantiGene Plex assay. This plate is a clear, polypropylene 96-well plate with round-bottom wells. Use with QuantiGene Plex Pressure Seals (Cat. No. QP0544) only.
Specifications
Specifications
| Barcode | No Barcode |
| Array | 8 x 12 |
| Product Line | QuantiGene Plex |
| Color | Clear |
| Product Type | PCR Microplate |
| Material | Polypropylene |
| No. of Wells | 96 |
| Well Shape | Round Bottom (U-well) |
| Quantity | 10 Plates |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.