Learn More
Invitrogen™ QuantiGene™ Plex Magnetic Separation Plates
Catalog No. QP0540
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Plates
Catalog No. Quantity
QP0540 10 Plates
1 options
Catalog No. QP0540 Supplier Invitrogen™ Supplier No. QP0540
Add to Cart
Add to Cart
QuantiGene Magnetic Separation Plates are 96-well flat-bottom plates used for the branched DNA and SAPE incubations on the second day of the QuantiGene Plex Assay.
QuantiGene Magnetic Separation Plates are 96-well flat-bottom plates used for the branched DNA and SAPE incubations on the second day of the QuantiGene Plex Assay. The Magnetic Separation Plates are compatible with the Hand-Held Magnetic Plate Washer (Cat. No. EPX-55555-000) and QuantiGene Plex Plate Seals (Cat. No. QG0524). These plates are one component of several included in the QuantiGene Plex Assay kit and are offered here as a supplement to the assay kit when more plates are needed.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Array 8 x 12
Product Line QuantiGene Plex
Product Type Magnetic Separation Plate
No. of Wells 96
Well Shape Flat Bottom (F-well)
Quantity 10 Plates
Shipping Condition Room Temperature
For Research Use Only. Not for use in diagnostic procedures.