Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Plex Mouse Neuroscience Panel, 15-plex

Catalog No. 5000153564
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
10 Plates
3 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-001-53564 10 Plates
50-001-53553 1 Plate
50-001-53533 3 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-001-53564 Supplier Invitrogen™ Supplier No. QGP315MSNEUROSCI
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The QuantiGene Plex Mouse Neuroscience Panel 15-plex, enables measurement of expression of relevant mouse genes associated with neuroscience and brain development through the analysis of 15 RNA targets and housekeeping genes in a single well using Luminex xMAP technology.

The QuantiGene Plex Mouse Neuroscience Panel, 15-plex, enables measurement of expression of relevant genes in mice associated with neuroscience and brain development through the analysis of 15 RNA targets and housekeeping genes in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. This panel contains key genes that are involved in dynamic processes regulating brain development and neurobiology.

Targets: Ahsg, Apoe, Bdnf, Cfh, Cntf, Fgf21, Gdnf, Gfap, Ncam1, Ngf, Nrgn, Uchl1, Gapdh, Hprt, Ppib

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide outstanding levels of validation and precision.

This panel is used in combination with the QuantiGene Plex Assay Kit, which contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates.

Features of QuantiGene Plex panels include:

  • True multiplexing—measure multiple genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Accommodation of difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • Standardized platform—uses 96-plate format compatible with Luminex 200, FLEXMAP 3D, and INTELLIFLEX systems (384-well format available on demand)
  • Simple workflow—employs an ELISA-like workflow with direct hybridization of transcripts to beads and transcript labeling (branched DNA signal amplification and PE readout)
  • Customize with validated targets—if alternative targets are needed in this panel, select from our large inventory of over 22,000 genes to create custom research panels
  • Rapid turnaround for fast delivery

Panel and signal amplification technology

QuantiGene Plex assays are hybridization-based assays for the direct quantitation of RNA directly from cell lysates, tissue homogenates, or purified RNA, and they enable the measurement of multiple targets in the same well via multiplexing. QuantiGene Plex panels contain two components: a target-specific probe set and magnetic capture beads. The target-specific probes are used to capture the RNA of interest, and branched DNA technology then amplifies the fluorescent signals, which are read with a Luminex xMAP instrument. The intensity of the fluorescent signal is directly proportional to the amount of target mRNA present in the sample. This allows for (relative) quantification of gene expression and, with this specific panel, enables human immune profiling. Detailed and step-by-step data analysis is facilitated by the QuantiGene Plex data analysis app.

Applications

QuantiGene Plex assay research applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency determination, prospective and/or retrospective analysis of clinical samples, microarray validation, predictive toxicology research, and translocation/fusion gene detection. In general, any multiplex gene expression analysis can be done with QuantiGene Plex assays, independent of species or sample type.

TRUSTED_SUSTAINABILITY

Specifications

Format 96-well
For Use With (Application) QuantiGene Gene Expression Assay
For Use With (Equipment) Luminex Instrument
Content And Storage Target-specific Probe Set and magnetic Capture Beads (premixed and ready to use)
Store Probe Set at -20°C and Capture Beads at 4°C (do not freeze!)
Product Line QuantiGene™ Plex
Product Type Multiplex Panel
Quantity 10 Plates
Shipping Condition Wet Ice
Sample Type Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Detection Method Fluorescence
Research Area Neurobiology
Species Mouse
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.