Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Sample Processing Kit, blood samples

Catalog No. QS0112
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Plates
2 Plates
5 x 10 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0112 5 x 10 Plates
QS0110 2 Plates
QS0111 10 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. QS0112 Supplier Invitrogen™ Supplier No. QS0112
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your whole blood or dried blood spot sample

What you get
The kit contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing whole blood from heparin, citrate, or EDTA tubes, PAXgene blood RNA tubes, Tempus™ blood RNA tubes, or from dried blood spots prepared from standard anticoagulant venous blood or finger sticks.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 5 x 50 mL Lysis Mixture, store at 15°C to 30°C
• 5 x 3.25 mL Proteinase K (50 μg/μL), store at -20°C

Format Kit
Sample Type Blood
Product Line QuantiGene
Product Type Sample Processing Kit
Quantity 5 x 10 Plates
Shipping Condition Blue Ice
Can samples in TRIzol solution be run in the QuantiGene Plex assay?

No, samples in TRIzol solution cannot be run in the QuantiGene Plex assay. However, RNA purified from these samples can be run in the QuantiGene Plex assay.

Can I use the QuantiGene Plex assay with bacteria?

Although the kits were not originally validated for bacteria, we have guidelines for preparing bacterial samples for QuantiGene Plex assays: http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/preparation-bacterial-homogenates-quantigene-plex-assays-protocol.pdf

Do you offer QuantiGene Plex assay custom services where I can send my samples for you to process?

Yes, please reach out to your Technical Sales Specialist for more information or fill out this form: https://www.thermofisher.com/us/en/home/global/forms/life-science/request-quote-quantigene-assays.html

Can I detect genomic DNA using the QuantiGene Plex assay?

Under the hybridization conditions that are used for the QuantiGene Plex assay, genomic DNA remains double-stranded. Therefore, it is not possible for the probes to bind to DNA. The signal from the probe sets comes only from RNA.

Can I use other lysis buffers with QuantiGene Plex assay samples?

We only recommend the use of QuantiGene Lysis Mixture for preparing cell and blood lysates. The Lysis Mixture has been optimized to provide stability of RNA in the lysates. In addition, QuantiGene Lysis Mixture provides the appropriate buffer conditions for the overnight hybridization.

Does Thermo Fisher Scientific currently perform functional testing on the QuantiGene Plex assay probe sets?

For QuantiGene Plex assays, we do perform functional QC of the probe sets using total RNA preparations for the species of interest.

How should I seal the Day 1 plate for Target Probe hybridization for the 384-well QuantiGene Plex assay?

The Day 1 plate can be sealed either manually or using a heat sealer, which is the preferred option. The pressure seal for manual sealing and the heat seal (Cat. No. AB-0757 for Thermo Fisher ALPS 50 V-Manual Heat Sealer, Cat. No. AB-1443A) are provided in the kit. If a different heat sealer will be used, please consult the manufacturer for suitable sealing.

Can I use a heat sealer to seal the plate on Day 2 of the QuantiGene Plex assay?

No, manual sealing using the provided seal is recommended. Heat sealing would deform the plate causing problem in washing.

Should I use control/housekeeping genes in my panel for QuantiGene Plex assays?

Three or more control/housekeeping genes are recommended to normalize the gene expression results with QuantiGene Plex assays. Please refer to the application note "Defining a QuantiGene Plex Assay panel" (https://assets.thermofisher.com/TFS-Assets/LSG/brochures/defining-quantigene-plex-assay-panel-app-note.pdf).

Can I use QuantiGene Plex assays for miRNA or DNA analysis?

miRNA and DNA can only be analyzed with our QuantiGene Singleplex assays, that allow for detection of one target per well.
Here is the link to the QuantiGene Singleplex website: https://www.thermofisher.com/us/en/home/life-science/gene-expression-analysis-genotyping/quantigene-rna-assays/quantigene-singleplex-assay.html

What reagents are included in the QuantiGene Plex assay kit?

The QuantiGene Plex assay kit provides all necessary reagents to perform the assay except Target Probe and Bead mix which are ordered separately. Please refer to the user guide for a list of the reagents included.

What are the proper storage conditions for the QuantiGene Plex assay reagents?

When the QuantiGene Plex assay kit arrives, please follow the instructions in the product sheet to store individual components. The Pre-Amp/Amp/LP Solutions, Bead Mix, SAPE, SAPE Diluent should be stored at 4 degrees C, while the probe set mix, Proteinase K, and blocking reagent should be stored at -20 degrees C. Other reagents should be stored at room temperature.

For QuantiGene Plex assays, what is the recommended temperature for hybridization?

We recommend using Day 1 hybridization temperature of 54 degrees C and Day 2 hybridization temperature of 50 degrees C. Please use QuantiGene Temperature Validation Kit (Cat. No. QS0517) to verify the incubator temperature.

What data analysis software solutions are offered for QuantiGene Plex assays?

The QuantiGene data analysis app is available on the Thermo Fisher Connect Platform for analyzing QuantiGene Plex assay data. For advanced QuantiGene Plex assay data analysis and visualization, the free desktop application software, Applied Biosystems Transcriptome Analysis Console (TAC) software can be used.

What sample types are compatible with QuantiGene Plex assays?

Whole blood, PaxGene blood, fresh tissue or FFPE tissues, cultured cells, and purified RNA have been tested with QuantiGene Plex assays. Other sample types including Tempus blood, bacteria, H&E stained sections, and blood spots are also compatible. Various sample prep protocols and kits are available.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.