Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Sample Processing Kit, cultured cells

Catalog No. QS0100
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Plates
2 Plates
5 x 10 Plates
50 Plates
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0100 2 Plates
QS0101 10 Plates
QS0103 50 Plates
QS0102 5 x 10 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. QS0100 Supplier Invitrogen™ Supplier No. QS0100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This QuantiGene Sample Processing Kit for cultured cells contains reagents and instructions for the preparation of cell lysates for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA targets and QuantiGene Plex DNA assays for DNA targets.

This QuantiGene Sample Processing Kit for cultured cells contains reagents and instructions for the preparation of cell lysates for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA targets and QuantiGene Plex DNA assays for DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your cultured cell sample

What you get
The QuantiGene Sample Processing Kit for cultured cells contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing cell lysates from cultured cells.

Success in cell lysis
Key factors that influence the complete lysis of the cells include:

  • The lysis mixture
  • Proteinase K
  • Vortexing or pipetting of the sample
  • Storage of the lysates at -80°C
  • Incubation at 50°C

The protocols provided with the QuantiGene Sample Processing Kit for cultured cells use a combination of these factors to ensure complete lysis of the samples. In addition, the lysis mixture provides the proper ionic conditions for the overnight hybridization in the QuantiGene assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 15 mL Lysis Mixture, store at 15°C to 30°C
• 150 μL Proteinase K (50 μg/μL), store at -20°C

Format Kit
Sample Type Cell Cultures
Product Line QuantiGene
Product Type Sample Processing Kit
Quantity 2 Plates
Shipping Condition Blue Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.