Learn More
Invitrogen™ QuantiGene™ Sample Processing Kit, tissues
Description
The QuantiGene Sample Processing Kit for tissues contains reagents and instructions for the preparation of tissue homogenates from fresh or frozen animal or plant tissues for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
- Easy to follow instructions
- Preserves the integrity of the RNA/DNA without degradation
- Choose the method that matches your tissue sample
What you get
The QuantiGene Sample Processing Kit for tissues contains our proprietary homogenizing solution, proteinase K, and detailed instructions for preparing homogenized tissue from a variety of tissue sources.
Specifications
Specifications
| Content And Storage | • 10 mL Homogenizing Solution, store at 15°C to 30°C • 36 μL Proteinase K (50 μg/μL), store at -20°C |
| Format | Kit |
| Sample Type | Tissue |
| Product Line | QuantiGene |
| Product Type | Sample Processing Kit |
| Quantity | 10 Samples |
| Shipping Condition | Blue Ice |
Frequently Asked Questions (FAQs)
No, samples in TRIzol solution cannot be run in the QuantiGene Plex assay. However, RNA purified from these samples can be run in the QuantiGene Plex assay.
Although the kits were not originally validated for bacteria, we have guidelines for preparing bacterial samples for QuantiGene Plex assays: http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/preparation-bacterial-homogenates-quantigene-plex-assays-protocol.pdf
Yes, please reach out to your Technical Sales Specialist for more information or fill out this form: https://www.thermofisher.com/us/en/home/global/forms/life-science/request-quote-quantigene-assays.html
Under the hybridization conditions that are used for the QuantiGene Plex assay, genomic DNA remains double-stranded. Therefore, it is not possible for the probes to bind to DNA. The signal from the probe sets comes only from RNA.
We only recommend the use of QuantiGene Lysis Mixture for preparing cell and blood lysates. The Lysis Mixture has been optimized to provide stability of RNA in the lysates. In addition, QuantiGene Lysis Mixture provides the appropriate buffer conditions for the overnight hybridization.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.