Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuantiGene™ Singleplex Assay Kit

Catalog No. p-7203099
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Plates
2 Plates
50 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0012 2 Plates
QS0013 10 Plates
QS0016 50 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. QS0012 Supplier Invitrogen Supplier No. QS0012
Only null left
Add to Cart
Add to Cart

QuantiGene Singleplex assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates, tissue homogenates or purified RNA.

QuantiGene Singleplex assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates, tissue homogenates or purified RNA. They utilize branched DNA technology for signal amplification, and signal is detected using a standard luminometer. The QuantiGene Singleplex Assay Kit contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates. Sample processing kits and target-specific probe sets are sold separately.

The QuantiGene Singleplex Assay is comprised of three modules:

  • QuantiGene Singleplex Assay Kit
  • QuantiGene Singleplex target-specific probe sets (sold separately)
  • QuantiGene Sample Processing Kit (sold separately) (not required for purified RNA samples)

Features of the QuantiGene Singleplex Assay include:

  • Direct from lysate quantitation using signal amplification—no RNA or DNA purification, reverse transcription, or PCR amplification needed
  • Precisely quantify subtle differences in gene expression as small as 10%
  • Simple workflow features lyse-and-go sample preparation, easily automatable ELISA-like protocol, and readout using any standard luminometer
  • Analyze any sample type: cultured cells; whole blood or dried blood spots; fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues; plants; bacteria

QuantiGene Singleplex assays bring the power of branched DNA (bDNA) technology to basic and pre-clinical research. The bDNA assay is a sandwich nucleic acid hybridization method that uses bDNA molecules to amplify signal from captured target RNA. bDNA technology forms the basis of the FDA-approved clinical diagnostic VERSANT 3.0 assays for HIV, HCV, and HBV viral load that are offered commercially by Siemens and have been in use for over a decade. Another advantage of bDNA assays is that RNA is measured directly from the sample source, without RNA purification or enzymatic manipulation, thereby avoiding inefficiencies and variability introduced by or errors inherent to these processes.

Each capture plate in this assay kit is packaged individually in a sealed foil pouch.

TRUSTED_SUSTAINABILITY

Specifications

Format 96-well
For Use With (Application) QuantiGene Singleplex Probe Set
For Use With (Equipment) Luminometer
Content And Storage

• Blocking Reagent (400 μL, store at 2°C to 8°C)
• Pre-Amplifier Solution (23 mL, store at 2°C to 8°C)
• Amplifier Solution (23 mL, store at 2°C to 8°C)
• Capture Plate (2 ea, packaged individually, store at 2°C to 8°C)
• Label Probe Solution (23 mL, store at 2°C to 8°C)
• Substrate (24 mL, store at 2°C to 8°C)
• Lysis Mixture (18 mL, store at 15–30°C)
• Wash Buffer Component 1 (3 mL, store at 15–30°C)
• Wash Buffer Component 2 (5 mL, store at 15–30°C)
• Plate Seals Day 1 (2 ea, store at 15–30°C)
• Plate Seals Day 2 (10 ea, store at 15–30°C)

Product Line QuantiGene Singleplex
Product Type Singleplex Assay Kit
Quantity 2 Plates
Detection Method Chemiluminescence

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.