Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Singleplex DNA, 1000 reactions

Catalog No. CQGSD1000
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CQGSD1000 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CQGSD1000 Supplier Invitrogen™ Supplier No. CQGSD1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a customizable DNA probe set for use with the QuantiGene Singleplex DNA Assay.

This is a customizable DNA probe set for use with the QuantiGene Singleplex DNA Assay.

TRUSTED_SUSTAINABILITY

Specifications

Product Line QuantiGene
Quantity 1000 reactions
Content And Storage QuantiGene Singleplex DNA probe set, 445 μL

Store at -20°C.
No. of Reactions 1000 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.