Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Singleplex DNA, 5000 reactions

Catalog No. CQGSD5000
Encompass_Preferred
Change view
Click to view available options
Quantity:
5000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CQGSD5000 5000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CQGSD5000 Supplier Invitrogen™ Supplier No. CQGSD5000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a customizable DNA probe set for use with the QuantiGene Singleplex DNA Assay.

This is a customizable DNA probe set for use with the QuantiGene Singleplex DNA Assay.

TRUSTED_SUSTAINABILITY

Specifications

Product Line QuantiGene
Quantity 5000 reactions
Content And Storage QuantiGene Singleplex DNA probe set, 2.2 mL

Store at -20°C.
No. of Reactions 5000 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.