Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuantiGene™ Singleplex HT Assay Kit

Catalog No. QS0384
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 x 384-well plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0384 50 x 384-well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QS0384 Supplier Invitrogen Supplier No. QS0384
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

QuantiGene™ Singleplex HT Assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates or tissue homogenates.

QuantiGene Singleplex HT assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates or tissue homogenates. They utilize branched DNA technology for signal amplification and signal is detected using a standard luminometer. The QuantiGene Singleplex Assay Kit contains all buffers, plates, and detection reagents necessary to perform the assay in 384-well plates.

Additional plate seals are available for separate purchase (Cat. No. QS0506).
TRUSTED_SUSTAINABILITY

Specifications

Format 384-well
For Use With (Application) QuantiGene Singleplex Probe Set
For Use With (Equipment) Luminometer
Content And Storage

• Blocking Reagent (2 x 10 mL, store at 2°C to 8°C)
• Pre-Amplifier Solution (3 x 400 mL, store at 2°C to 8°C)
• Amplifier Solution (3 x 400 mL, store at 2°C to 8°C)
• Capture Plate (50 ea, packaged individually, store at 2°C to 8°C)
• Label Probe Solution (3 x 400 mL, store at 2°C to 8°C)
• Substrate (4 x 300 mL, store at 2°C to 8°C)
• Lysis Mixture (510 mL, store at 15–30°C)
• Wash Buffer Component 1 (2 x 75 mL, store at 15–30°C)
• Wash Buffer Component 2 (2 x 125 mL, store at 15–30°C)
• Plate Seals (260 ea, store at 15–30°C)

Product Line QuantiGene Singleplex
Product Type Singleplex HT Assay Kit
Quantity 50 x 384-well plates
Shipping Condition Blue Ice
Sample Type Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Species Universal

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.