Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Singleplex RNA, 5000 reactions

Catalog No. CQGS5000
Encompass_Preferred
Change view
Click to view available options
Quantity:
5000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CQGS5000 5000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CQGS5000 Supplier Invitrogen™ Supplier No. CQGS5000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a customizable probe set for the QuantiGene Singleplex RNA Assay.

This is a customizable probe set for the QuantiGene Singleplex RNA Assay.

TRUSTED_SUSTAINABILITY

Specifications

Product Line QuantiGene
Quantity 5000 reactions
Content And Storage QuantiGene Singleplex RNA probe set, 3.9 mL

Store at -20°C.
No. of Reactions 5000 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.