Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Quantikine Immunoassay Control Group 3
SDP

Catalog No. QC20
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC20 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QC20 Supplier R&D Systems™ Supplier No. QC20

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Control for Human CCL3/MIP-1 alpha, CCL4/MIP-1 beta, CCL5/RANTES, CD14, CNTF, CXCL12/SDF-1 alpha, FGF basic, Follistatin, IL-5, IL-13, PDGF-AB, SCF, TGF-beta 2, and VEGF-D

  • For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems' Quantikine™ kits
  • Controls are prepared in diluted porcine serum with preservatives
  • They contain recombinant human cytokines at low, medium and high concentrations
  • Controls are supplied lyophilized

Specifications

Product Type Quantikine Immunoassay Control Group 3
For Use With (Application) Control
Quantity 1 Each
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.