Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 1099 Human S100A8/S100A9
SDP

Catalog No. QC223
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC223 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QC223 Supplier R&D Systems™ Supplier No. QC223

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with catalog number DS8900

Long Name: S100 Calcium Binding Protein A8/S100 Calcium Binding Protein A9

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Quantikine Immunoassay Control Set 893 Human S100A8/S100A9
Gene ID (Entrez) 6279 (Human)
Intended Use Control
Quantity 1 Each
Source Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.