Learn More
ATCC Quantitated genomic DNA; from Cowpox virus; Strain Brighton
Catalog No. 502382245
Encompass
Quantitative genomic DNA from Cowpox virus strain Brighton can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2245 Supplier ATCC Supplier No. VR302DQ
Quantitative genomic DNA from Cowpox virus strain Brighton can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.