Learn More
ATCC Quantitative Genomic; DNA from Pseudomonas; paraeruginosa; Strain R. Hugh 813
Supplier:  ATCC 9027DQ
Encompass
Quantitative genomic DNA from Pseudomonas paraeruginosa strain R. Hugh 813 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-2879