Learn More
ATCC Quantitative Genomic DNA from; Human adenovirus 1; Strain: Adenoid 71
Catalog No. 502384894
Encompass
Quantitative genomic DNA from Human adenovirus 1 strain Adenoid 71 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4894 Supplier ATCC Supplier No. VR1DQ
Quantitative genomic DNA from Human adenovirus 1 strain Adenoid 71 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.