Learn More
ATCC Quantitative Genomic DNA from; Canid herpesvirus 1; Strain: D 004
Catalog No. 502384877
Encompass
Quantitative Genomic DNA from Canid herpesvirus 1 strain D 004 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4877 Supplier ATCC Supplier No. VR552DQ
Quantitative Genomic DNA from Canid herpesvirus 1 strain D 004 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.