Learn More
ATCC Quantitative Genomic DNA; from Legionella pneumophila; subsp. pneumophila; Strain: Philadelphia-1
Catalog No. 502383914
Encompass
Quantitative genomic DNA from Legionella pneumophila subsp. pneumophila strain Philadelphia-1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3914 Supplier ATCC Supplier No. 502383914
Quantitative genomic DNA from Legionella pneumophila subsp. pneumophila strain Philadelphia-1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.