Learn More
ATCC Quantitative Genomic DNA from; Klebsiella pneumoniae; Strain: ART 2008133
Catalog No. 502383029
Encompass
Quantitative Genomic DNA from Klebsiella pneumoniae strain ART 2008133 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3029 Supplier ATCC Supplier No. BAA1705DQ
Quantitative Genomic DNA from Klebsiella pneumoniae strain ART 2008133 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.