Learn More
ATCC Quantitative Genomic DNA from; Aspergillus fumigatus; Strain: 118
Supplier:  ATCC 1022DQ
Encompass
Quantitative Genomic DNA from Aspergillus fumigatus strain NRRL 163 can be used for assay development, verification, validation, monitoring day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-238-3180