Learn More
ATCC Quantitative Genomic DNA from; Aspergillus flavus; Strain: SN 3
Catalog No. 502383607
Encompass
Quantitative Genomic DNA from Asperigillus flavus strain SN 3 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-238-3607 Supplier ATCC Supplier No. 9643DQ
Quantitative Genomic DNA from Asperigillus flavus strain SN 3 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.