Learn More
ATCC Quantitative Genomic DNA from; Clostridium perfringens; Strain: NCTC 8237
Catalog No. 502381843
Encompass
Quantitative genomic DNA from Clostridium perfringens strain NCTC 8237 (ATCC 13124) can be used for assay development, verification, validation, and monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-1843 Supplier ATCC Supplier No. 13124DQ
Quantitative genomic DNA from Clostridium perfringens strain NCTC 8237 (ATCC 13124) can be used for assay development, verification, validation, and monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.