Learn More
ATCC Quantitative Genomic DNA from; Candida glabrata; Strain: CBS 138
Catalog No. 502382463
Encompass
Quantitative Genomic DNA from Candida glabrata strain CBS 138 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-238-2463 Supplier ATCC Supplier No. 2001DQ
Quantitative Genomic DNA from Candida glabrata strain CBS 138 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.