Learn More
ATCC Quantitative Genomic DNA from; Cereibacter sphaeroides; Strain: ATH 2.4.1
Catalog No. 502383056
Encompass
Quantitative genomic DNA from Cereibacter sphaeroides strain ATH 2.4.1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3056 Supplier ATCC Supplier No. 17023DQ
Quantitative genomic DNA from Cereibacter sphaeroides strain ATH 2.4.1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.