Learn More
ATCC Quantitative Genomic DNA from; Aspergillus niger; Strain: A1144 3528.7
Catalog No. 502383177
Encompass
Quantitative Genomic DNA from Aspergillus niger strain A1144 3528.7 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-238-3177 Supplier ATCC Supplier No. 1015DQ
Quantitative Genomic DNA from Aspergillus niger strain A1144 3528.7 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.