Learn More
ATCC Quantitative Genomic DNA from; Streptococcus pneumoniae; Strain: 262
Catalog No. 502383366
Encompass
Quantitative Genomic DNA from Streptococcus pneumoniae strain 262 [CIP 104340] can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3366 Supplier ATCC Supplier No. 49619DQ
Quantitative Genomic DNA from Streptococcus pneumoniae strain 262 [CIP 104340] can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.