Learn More
ATCC Quantitative Genomic DNA from; Human adenovirus 4; Strain: RI-67
Supplier:  ATCC VR1572DQ
Encompass
Quantitative Genomic DNA from Human adenovirus 4 strain RI-67 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2073