Learn More
ATCC Quantitative Genomic DNA from; Enterococcus faecalis; NJ-3
Catalog No. 502382149
Encompass
Quantitative Genomic DNA from Enterococcus faecalis strain NJ-3 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-2149 Supplier ATCC Supplier No. 51299DQ
Add to Cart
Add to Cart
Quantitative Genomic DNA from Enterococcus faecalis strain NJ-3 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.