Learn More
ATCC Quantitative Genomic DNA from; Campylobacter coli; Strain: CIP 7080
Catalog No. 502382750
Encompass
Quantitative genomic DNA from Campylobacter coli strain CIP 7080 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-2750 Supplier ATCC Supplier No. 33559DQ
Quantitative genomic DNA from Campylobacter coli strain CIP 7080 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.