Learn More
ATCC Quantitative Genomic DNA from; Salmonella enterica; subsp. enterica; serovar Typhimurium
Catalog No. 502383485
Encompass
Quantitative Genomic DNA from Salmonella enterica subsp. enterica serovar Typhimurium strain CDC 6516-60 can be used for assay development, verification and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3485 Supplier ATCC Supplier No. 14028DQ
Quantitative Genomic DNA from Salmonella enterica subsp. enterica serovar Typhimurium strain CDC 6516-60 can be used for assay development, verification and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.