Learn More
ATCC Quantitative Genomic DNA from; Human adenovirus 3; Strain: GB
Catalog No. 502381721
Encompass
Genomic DNA isolated from a preparation of Human adenovirus 3 strain GB (ATCC VR-3). This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-1721 Supplier ATCC Supplier No. VR3DQ
Genomic DNA isolated from a preparation of Human adenovirus 3 strain GB (ATCC VR-3). This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.