Learn More
ATCC Quantitative Genomic DNA from; Klebsiella pneumoniae; Strain: NCTC 9633
Catalog No. 502383805
Encompass
Quantitative Genomic DNA from Klebsiella pneumoniae subsp. pneumoniae strain NCTC 9633 (ATCC 13883) can be used for assay development, verification, and validation as well as monitoring day-to-day test variation and the lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3805 Supplier ATCC Supplier No. 13883DQ
Quantitative Genomic DNA from Klebsiella pneumoniae subsp. pneumoniae strain NCTC 9633 (ATCC 13883) can be used for assay development, verification, and validation as well as monitoring day-to-day test variation and the lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.