Learn More
ATCC Quantitative Genomic DNA from; Staphylococcus saprophyticus; subsp. saprophyticus
Catalog No. 502384004
Encompass
Quantitative genomic DNA from Staphylococcus saprophyticus subsp. saprophyticus strain NCTC 7292 (ATCC 15305) can be used for assay development, verification, and validation as well as monitoring day-to-day test variation and the lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4004 Supplier ATCC Supplier No. 15305DQ
Quantitative genomic DNA from Staphylococcus saprophyticus subsp. saprophyticus strain NCTC 7292 (ATCC 15305) can be used for assay development, verification, and validation as well as monitoring day-to-day test variation and the lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.