Learn More
ATCC Quantitative Genomic DNA from; Human adenovirus 2; Strain: Adenoid 6
Catalog No. 502384047
Encompass
Quantitative genomic DNA from Human adenovirus 2 strain Adenoid 6 can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4047 Supplier ATCC Supplier No. VR846DQ
Quantitative genomic DNA from Human adenovirus 2 strain Adenoid 6 can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.