Learn More
ATCC Quantitative Genomic DNA from; Micrococcus luteus; Strain: 130.21
Catalog No. 502384199
Encompass
Genomic DNA isolated from a preparation of Micrococcus luteus strain 130.21 (ATCC 10240). This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4199 Supplier ATCC Supplier No. 10240DQ
Genomic DNA isolated from a preparation of Micrococcus luteus strain 130.21 (ATCC 10240). This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.