Learn More
ATCC Quantitative Genomic DNA from; Enterococcus faecalis; Strain: [Portland]
Catalog No. 502381735
Encompass
Quantitative Genomic DNA from Enterococcus faecalis can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-1735 Supplier ATCC Supplier No. 29212DQ
Quantitative Genomic DNA from Enterococcus faecalis can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.