Learn More
ATCC Quantitative Genomic DNA; from Haemophilus influenzae; Strain Rd [KW20]
Catalog No. 502384575
Encompass
Genomic DNA from Haemophilus influenzae strain Rd can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4575 Supplier ATCC Supplier No. 51907DQ
Genomic DNA from Haemophilus influenzae strain Rd can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.