Learn More
ATCC Quantitative Genomic DNA from; Klebsiella quasipneumoniae; Strain: K6
Catalog No. 502384898
Encompass
Genomic DNA isolated from a preparation of Klebsiella quasipneumoniae strain K6 (ATCC 700603). This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4898 Supplier ATCC Supplier No. 700603DQ
Genomic DNA isolated from a preparation of Klebsiella quasipneumoniae strain K6 (ATCC 700603). This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.